Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse Wnt-5a Biotinylated Protein

Catalog No. bt645010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human/Mouse Wnt-5a Biotinylated Protein is derived from CHO. The Recombinant Human/Mouse Wnt-5a Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P22725
For Use With (Application) Bioactivity
Gene ID (Entrez) 7474
Molecular Weight (g/mol) 38 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived Wnt-5a protein Gln38-Lys380
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity Wnt-5a
Conjugate Unconjugated
Species Human, Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS, EDTA, CHAPS and Trehalose with BSA as a carrier protein.
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.