Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MRPS28 His Protein
SDP

Catalog No. NBP25194301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25194301 0.1 mg
NBP25194305 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25194301 Supplier Novus Biologicals™ Supplier No. NBP2519430.1MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 72-187 of Human MRPS28 The Recombinant Human MRPS28 Protein is derived from E. coli. The Recombinant Human MRPS28 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mMPhosphate buffer saline (pH 8.0) containing 20% glycerol, 1mM DTT
Gene ID (Entrez) 28957
Molecular Weight (g/mol) TMW: 15.5kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias 28S ribosomal protein S35, mitochondrial, FLJ22853, mitochondrial 28S ribosomal protein S35, mitochondrial ribosomal protein S28, MRP-S2828S ribosomal protein S28, mitochondrial, MRP-S35, MRPS35HSPC007, S28mt, S35mt
Common Name MRPS28
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.