Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MST2/STK3 Protein
SDP

Catalog No. NB253049250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253049250 250 μg
NB25304950 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253049250 Supplier Novus Biologicals™ Supplier No. NBP253049250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-322 of Human MST2/STK3 The Recombinant Human MST2/STK3 Protein is derived from E. coli. The Recombinant Human MST2/STK3 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 6788
Molecular Weight (g/mol) 37.6kDa
Name Human MST2/STK3 Protein
Purification Method >90%, by SDS-PAGE
Quantity 250 μg
Source Baculovirus
Immunogen ADPMEQPPAP KSKLKKLSED SLTKQPEEVF DVLEKLGEGS YGSVFKAIHK ESGQVVAIKQ VPVESDLQEI IKEISIMQQC DSPYVVKYYG SYFKNTDLWI VMEYCGAGSV SDIIRLRNKT LIEDEIATIL KSTLKGLEYL HFMRKIHRDI KAGNILLNTE GHAKLADFGV AGQLTDTMAK RNTVIGTPFW MAPEVIQEIG YNCVADIWSL GITSIEMAEG KPPYADIHPM RAIFMIPTNP PPTFRKPELW SDDFTDFVKK CLVKNPEQRA TATQLLQHPF IKNAKPVSIL RDLITEAMEI KAKRHEEQQR ELEEEEENSD EDELDHHHHH H
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2
Gene Symbol STK3
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.