Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Myoglobin Protein
SDP

Catalog No. NB398101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398101 500 μg
NB398102 1 mg
NB398103 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB398101 Supplier Novus Biologicals™ Supplier No. NBP307108500UG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Formulation 0.2 μm filtered solution in 25 mM Tris-HCl (pH 8.0)
Gene ID (Entrez) 4151
Molecular Weight (g/mol) MolecularWeight-theroretical: 17.1 kDa
Quantity 500 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1.0 EU/μg of Myoglobin as determined by LAL method.
Gene Alias MGC13548, myoglobin, PVALB
Gene Symbol MB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Cancer, Cardiovascular Biology
Purity or Quality Grade >95%, by SDS-PAGE
Protein Myoglobin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.