Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NEIL1 His Protein
SDP

Catalog No. NBP17884610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17884610 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17884610 Supplier Novus Biologicals™ Supplier No. NBP17884610UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-390 of Human NEIL1 The Recombinant Human NEIL1 Protein is derived from E. coli. The Recombinant Human NEIL1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q96FI4
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 0.1M NaCl,1mM DTT, 0.1mM PMSF
Gene ID (Entrez) 79661
Molecular Weight (g/mol) 45.8kDa
Name NEIL1 Protein
Quantity 10 μg
Source E. Coli
Immunogen NEIL1, 1-390aa, Human, His tag
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.