Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human NM23-H1 Protein

Click to view available options
:
0.1mg
Description
An un-tagged recombinant protein corresponding to the amino acids 1-152 of Human NM23-H1 The Recombinant Human NM23-H1 Protein is derived from E. coli. The Recombinant Human NM23-H1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | P15531 |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH7.5), 1mM DTT, 10% glycerol |
Gene ID (Entrez) | 4830 |
Molecular Weight (g/mol) | 17.1kDa |
Name | NM23-H1 Protein |
Purification Method | Protein |
Quantity | 0.1mg |
Source | E. Coli |
Immunogen | NME1, 1-152 aa, Human, Recombinant, E.coli (NP_000260). |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction