Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NM23-H2/NME2 Protein
SDP

Catalog No. NBP140409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140409 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140409 Supplier Novus Biologicals™ Supplier No. NBP140409
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-152 of Human NM23-H2/NME2 The Recombinant Human NM23-H2/NME2 Protein is derived from E. coli. The Recombinant Human NM23-H2/NME2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P22392
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 4831
Molecular Weight (g/mol) 17.2kDa
Name NM23-H2/NME2 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen NME2, 1-152aa, Human, Recombinant, E. coli (NP_001018149).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.