Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Noggin Protein
SDP

Catalog No. NB398116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398116 5 μg
NB398119 100 μg
NB398118 1 mg
NB398117 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398116 Supplier Novus Biologicals™ Supplier No. NBP3071045UG

Recombinant Protein

Biological activity measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.04-0.2 μg/mL in the presence of 50 ng/mL of Recombinant Human BMP-4.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS (pH 7.4), 0.02% Tween-20.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) MolecularWeight-theroretical: 23 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of Noggin as determined by LAL method.
Gene Alias noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Gene Symbol NOG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein Noggin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.