Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NPM1 Protein
SDP

Catalog No. N275952100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N275952100U 100 μg
N27595205M 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N275952100U Supplier Novus Biologicals™ Supplier No. NBP275952100UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-294 of Human NPM1

The Recombinant Human NPM1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 4869
Molecular Weight (g/mol) 34.7 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol NPM1
Specific Reactivity NPM1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 20 mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.15 M NaCl, 1 mM DTT
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.