Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human NSDHL His Protein
SDP

Catalog No. NB170427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170427 0.05 mg
NBP223258 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170427 Supplier Novus Biologicals™ Supplier No. NBP2232580.05MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-297 of Human NSDHL The Recombinant Human NSDHL Protein is derived from E. coli. The Recombinant Human NSDHL Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_057006
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 50814
Molecular Weight (g/mol) 35.5 kDa
Name NSDHL Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen NSDHL, 1-297 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMEPAVSE PMRDQVARTH LTEDTPKVNA DIEKVNQNQA KRCTVIGGSG FLGQHMVEQL LARGYAVNVF DIQQGFDNPQ VRFFLGDLCS RQDLYPALKG VNTVFHCASP PPSSNNKELF YRVNYIGTKN VIETCKEAGV QKLILTSSAS VIFEGVDIKN GTEDLPYAMK PIDYYTETKI LQERAVLGAN DPEKNFLTTA IRPHGIFGPR DPQLVPILIE AARNGKMKFV IGNGKNLVDF TFVENVVHGH ILAAEQLSRD STLGGKAFHI TNDEPIPFWT FLSRILTGLN YEAPKYHIPY
Storage Requirements -20°Cor to +80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.