Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | Lyophilized from a 0.2 micron filtered solution, 0.1% Trifluoroacetic Acid (TFA) |
| Gene ID (Entrez) | 4908 |
| Quantity | 500 μg |
| Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
| Gene Alias | HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3 |
| Gene Symbol | NTF3 |
| Reconstitution | Sterile water at 0.1 mg/mL |
| Research Category | Apoptosis, Cancer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
