Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human NT-3 Protein

Click to view available options
Quantity:
1 mg
100 μg
500 μg
Description
Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), >5.0 x 10^4 units/mg (ED50 acceptance criteria are <20 ng/mL).
Specifications
Specifications
For Use With (Application) | Bioactivity, SDS-PAGE |
Formulation | Lyophilized from a 0.2 micron filtered solution, 0.1% Trifluoroacetic Acid (TFA) |
Gene ID (Entrez) | 4908 |
Quantity | 500 μg |
Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
Gene Alias | HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3 |
Gene Symbol | NTF3 |
Reconstitution | Sterile water at 0.1 mg/mL |
Research Category | Apoptosis, Cancer |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction