Learn More
R&D Systems™ Recombinant Human PA28 Activator alpha Subunit Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Quantity | 100 μg |
| Species | Human |
| Product Line | R&D Systems |
| Content And Storage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.