Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PARP Protein
SDP

Catalog No. NB002055 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002055 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002055 Supplier Novus Biologicals™ Supplier No. NBP19963550UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein for Human PARP The Recombinant Human PARP Protein is derived from Baculovirus. The Recombinant Human PARP Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized.
Gene ID (Entrez) 142
Molecular Weight (g/mol) 116 kDa
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1
Gene Symbol PARP1
Reconstitution Centrifuge the vial prior to opening. Add 100 μL dH2O for a concentration of 0.5 mg/mL. Final buffer concentration: 20 mM Tris (pH 8), 0.3 M NaCl, 0.1 mM EDTA, 1 mM DTT, plus lyophilization stabilizers.
Buffer Lyophilized.
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.