Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PDGF-AA Protein
SDP

Catalog No. NB398127 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398127 100 μg
NB398126 10 μg
NB398125 1 mg
NB398124 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398127 Supplier Novus Biologicals™ Supplier No. NBP307102100UG
Add to Cart
Add to Cart

Recombinant Protein

Biological activity measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. The ED50 for tis effect is 50-200 ng/mL in a fluorometric assay using a redox sensitive dye.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, 300 mM NaCl, pH 6.0.
Gene ID (Entrez) 5154
Molecular Weight (g/mol) MolecularWeight-theroretical: 28.8 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of PDGF-AA as determined by LAL method.
Gene Alias PDGF1, PDGF-A
Gene Symbol PDGFA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE
Protein PDGF-AA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.