Learn More
Description
The Recombinant Human PDGF-AB Heterodimer Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Gene ID (Entrez) | 5154 |
| Molecular Weight (g/mol) | 25.5 kDa |
| Purification Method | >98%, by SDS-PAGE |
| Quantity | 10 μg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | PDGFA |
| Reconstitution | Reconstitute in 10 mM acetic acid to 0.1 - 1.0 mg/mL. This solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
