Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PDGFA Protein
SDP

Catalog No. NB19984710U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB19984710U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB19984710U Supplier Novus Biologicals™ Supplier No. NBP19984710UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Recombinant bioactive protein containing 250 amino acids for Human PDGFA
  • The Recombinant Human PDGFA Protein is derived from E. coli
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 5154
Molecular Weight (g/mol) 28.5 kDa
Purification Method >98%, by SDS-PAGE
Quantity 10 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 ng/μg
Gene Symbol PDGFA
Reconstitution Reconstitute in 10 mM acetic acid to 0.1 - 1.0 mg/mL. This solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized with no additives.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.