Learn More
Description
Specifications
Specifications
| Accession Number | NP_003759 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 7.5), 1mM DTT, 10% glycerol |
| Gene ID (Entrez) | 8682 |
| Molecular Weight (g/mol) | 15kDa |
| Name | PEA-15 Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MAEYGTLLQD LTNNITLEDL EQLKSACKED IPSEKSEEIT TGSAWFSFLE SHNKLDKDNL SYIEHIFEIS RRPDLLTMVV DYRTRVLKIS EEDELDTKLT RIPSAKKYKD IIRQPSEEEI IKLAPPPKKA |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
