Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Perilipin-2/ADFP Protein
SDP

Catalog No. NB253039100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253039100 100 μg
NB25303905 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253039100 Supplier Novus Biologicals™ Supplier No. NBP253039100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-437 of Human Perilipin-2/ADFP The Recombinant Human Perilipin-2/ADFP Protein is derived from E. coli. The Recombinant Human Perilipin-2/ADFP Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M urea, 10% glycerol
Gene ID (Entrez) 123
Molecular Weight (g/mol) 49.3kDa
Name Human Perilipin-2/ADFP Protein
Purification Method >85%, by SDS-PAGE
Quantity 100 μg
Source E.Coli
Immunogen MASMTGGQQM GRGSMASVAV DPQPSVVTRV VNLPLVSSTY DLMSSAYLST KDQYPYLKSV CEMAENGVKT ITSVAMTSAL PIIQKLEPQI AVANTYACKG LDRIEERLPI LNQPSTQIVA NAKGAVTGAK DAVTTTVTGA KDSVASTITG VMDKTKGAVT GSVEKTKSVV SGSINTVLGS RMMQLVSSGV ENALTKSELL VEQYLPLTEE ELEKEAKKVE GFDLVQKPSY YVRLGSLSTK LHSRAYQQAL SRVKEAKQKS QQTISQLHST VHLIEFARKN VYSANQKIQD AQDKLYLSWV EWKRSIGYDD TDESHCAEHI ESRTLAIARN LTQQLQTTCH TLLSNIQGVP QNIQDQAKHM GVMAGDIYSV FRNAASFKEV SDSLLTSSKG QLQKMKESLD DVMDYLVDNT PLNWLVGPFY PQLTESQNAQ DQGAEMDKSS QETQRSEHKT H
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Gene Symbol PLIN2
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.