Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Phospho-Ubiquitin (S65) Protein

Catalog No. U102150
Click to view available options
Quantity:
150 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Phospho-Ubiquitin (S65) Protein is derived from E. coli. The Recombinant Human Phospho-Ubiquitin (S65) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P0CG47.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES.
Gene ID (Entrez) 6233
Molecular Weight (g/mol) Theoretical M.W.: 8.6kDa
Quantity 150 μg
Source E. coli-derived human Ubiquitin protein Met1 - Gly76 Phosphorylated on Ser 65.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Gene Alias RPS27A, UBA52, UBB, UBB ubiquitin B, UBC
Common Name Ubiquitin
Gene Symbol UBB
Biological Activity Reaction conditions will need to be optimized for each specific application.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Phosphorylated on Ser 65.
Purity or Quality Grade >98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.