Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PHPT1 Protein
SDP

Catalog No. NBP259581X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259581X 100 μg
NBP25958120 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259581X Supplier Novus Biologicals™ Supplier No. NBP259581100UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-125 of Human PHPT1 The Recombinant Human PHPT1 Protein is derived from E. coli. The Recombinant Human PHPT1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid. 20 mM Tris-HCl buffer (pH8.0) containing 0.2M NaCl, 2mM DTT, 10% glycerol
Gene ID (Entrez) 29085
Molecular Weight (g/mol) 15.9 kDa
Name Recombinant Human PHPT1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein jan
Gene Symbol PHPT1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.