Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PLGF His Protein
SDP

Catalog No. NB151686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
0.02 mg
0.05 mg
0.1 mg
0.25 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151686 0.01 mg
NB151687 0.02 mg
NB151688 0.05 mg
NB151689 0.1 mg
NB151690 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. NB151686 Supplier Novus Biologicals™ Supplier No. NBP3118470.01MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Specifications

Concentration 0.25 mg/ml
For Use With (Application) Functional Assay
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 5228
Molecular Weight (g/mol) 18.3 kDa
Quantity 0.01 mg
Storage Requirements Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration < 1 EU per 1ug of protein (determined by LAL method)
Gene Alias D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Gene Symbol PGF
Research Category Angiogenesis
Purity or Quality Grade >95%, by SDS-PAGE
Protein PLGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.