Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PP1 alpha/PPP1A His Protein
SDP

Catalog No. NBP17241850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17241850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17241850 Supplier Novus Biologicals™ Supplier No. NBP17241850UG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-330 of Human PP1 alpha/PPP1A The Recombinant Human PP1 alpha/PPP1A Protein is derived from E. coli. The Recombinant Human PP1 alpha/PPP1A Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number NP_002699
For Use With (Application) Blocking/Neutralizing, SDS-PAGE
Formulation Liquid. 50mM Tris-HCl buffer (pH8.0) containing 40% glycerol, 0.15M NaCl, 1mM DTT, 0.1mM PMSF, 1mM MnCl2
Gene ID (Entrez) 5499
Molecular Weight (g/mol) 39.7kDa
Name PP1 alpha/PPP1A Protein
Quantity 50 μg
Source E. Coli
Immunogen 1-330aa, Human, His tag
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.