Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Protein Kinase D2 Protein
SDP

Catalog No. NB002030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002030 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002030 Supplier Novus Biologicals™ Supplier No. NBP1994675UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant Human Protein Kinase D2 with N-terminal GST tag The Recombinant Human Protein Kinase D2 Protein is derived from Sf 9 (baculovirus). The Recombinant Human Protein Kinase D2 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 0.1 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol
Gene ID (Entrez) 25865
Molecular Weight (g/mol) 130 kDa
Quantity 5 μg
Source Sf 9 (baculovirus)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias DKFZP586E0820, EC 2.7.11, EC 2.7.11.13, nPKC-D2, PKD2HSPC187, protein kinase D2, serine/threonine-protein kinase D2
Gene Symbol PRKD2
Buffer 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol
Purity or Quality Grade >80% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.