Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Protein Phosphatase 1C gamma Protein
SDP

Catalog No. NBP2595825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2595825 5 μg
NBP25958220 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2595825 Supplier Novus Biologicals™ Supplier No. NBP2595825UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-323 of Human Protein Phosphatase 1C gamma The Recombinant Human Protein Phosphatase 1C gamma Protein is derived from E. coli. The Recombinant Human Protein Phosphatase 1C gamma Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.25mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 50% glycerol, 0.2M NaCl, 2mM DTT
Gene ID (Entrez) 5501
Molecular Weight (g/mol) 39.1 kDa
Name Recombinant Human Protein Phosphatase 1C gamma Protein
Purification Method >90%, by SDS-PAGE
Quantity 5 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein
Gene Symbol PPP1CC
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.