Learn More
R&D Systems™ Recombinant Human PVRIG Fc Chimera Protein
Description
Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. It is composed of single extracellular IgV domain, one transmembrane domain, and long intracellular domain. The intracellular domain contains two tyrosine residues, one within an ITIM-like motif that is potential docking site for phosphatases. Reconstitute at 500μg/mL in PBS
Storage:
Store the unopened product at −20 to −70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Accession Number | Q6DKI7 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and PEG with Trehalose. |
Gene ID (Entrez) | 79037 |
Molecular Weight (g/mol) | 40 kDa |
Quantity | 50 μg |
Source | Human embryonic kidney cell, HEK293-derived human PVRIG protein, (N-terminus) Human PVRIG (Thr41 & Glu43-Leu172) Accession # Q6DKI7 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
Regulatory Status | RUO |
Structural Form | Disulfide-linked homodimer |
Show More |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.