Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human QTRTD1 His Protein
SDP

Catalog No. NBP25187825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25187825 0.25 mg
NBP25187805 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25187825 Supplier Novus Biologicals™ Supplier No. NBP2518780.25MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-415 of Human QTRTD1 The Recombinant Human QTRTD1 Protein is derived from E. coli. The Recombinant Human QTRTD1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffer saline (pH 7.4) containing 30% glycerol, 2mM DTT, 1mM EDTA, 0.1mM PMSF
Gene ID (Entrez) 79691
Molecular Weight (g/mol) TMW: 49.1kDa
Quantity 0.25 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 2.4.2.29, FLJ12960, queuine tRNA-ribosyltransferase domain containing 1, Queuine tRNA-ribosyltransferase domain-containing protein 1, queuine tRNA-ribosyltransferase subunit QTRTD1
Common Name QTRTD1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.