Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RAP30 His Protein
SDP

Catalog No. NB170707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170707 0.5 mg
NB170708 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170707 Supplier Novus Biologicals™ Supplier No. NBP3179830.5MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.2 M NaCl, 1 mM DTT
Gene ID (Entrez) 2963
Molecular Weight (g/mol) M.W. Theoretical: 30.5 kDa
Quantity 0.5 mg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias ATP-dependent helicase GTF2F2, BTF4, General transcription factor IIF 30 kDa subunit, general transcription factor IIF subunit 2, general transcription factor IIF, polypeptide 2, 30kDa, TF2F2, TFIIF, TFIIF-beta, Transcription initiation factor IIF subunit beta, Transcription initiation factor RAP30
Gene Symbol GTF2F2
Research Category Chromatin Research
Purity or Quality Grade >90%, by SDS-PAGE
Protein RAP30
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.