Learn More
Description
Specifications
Specifications
| Accession Number | O43566 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer, pH7.5, 10% glycerol, 1mM DTT, 200mM NaCl |
| Gene ID (Entrez) | 10636 |
| Name | RGS14 Protein |
| Purification Method | SDS-PAGE |
| Quantity | 20 μg |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >80% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
