Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Ribonuclease A His Protein
SDP

Catalog No. NB170677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170677 10 μg
NB170681 50 μg
NB170680 500 μg
NB170679 250 μg
NB170678 20 μg
NB170676 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. NB170677 Supplier Novus Biologicals™ Supplier No. NBP31216410UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 3 X 10^6 unit/mg, and is defined as the amount of enzyme that cleaves 1.0 nmole of RNA per minute at 25C.

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 6035
Molecular Weight (g/mol) M.W. Theoretical: 15.3 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1
Gene Symbol RNASE1
Research Category DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
Purity or Quality Grade >90%, by SDS-PAGE
Protein Ribonuclease A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.