Learn More
Description
Specifications
Specifications
| Accession Number | O43353 |
| Formulation | Recombinant proteins in storage buffer (50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol). |
| Gene ID (Entrez) | 8767 |
| Name | RIPK2 Protein |
| Quantity | 0.005 mg |
| Source | Baculovirus (Sf9 insect cells) |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
