Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human S100A8 Protein
SDP

Catalog No. NBP144482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144482 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144482 Supplier Novus Biologicals™ Supplier No. NBP144482
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to the amino acids 1- 93 of Human S100A8 The Recombinant Human S100A8 Protein is derived from E. coli. The Recombinant Human S100A8 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P05109
For Use With (Application) ELISA, In vivo Assay, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 6279
Molecular Weight (g/mol) 10.8kDa
Name S100A8 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen S100A8, 1- 93 aa, Human, Recombinant, E.coli (NP_002955).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.