Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human S100B Protein
SDP

Catalog No. NB398108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398108 5 μg
NB398111 100 μg
NB398110 1 mg
NB398109 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398108 Supplier Novus Biologicals™ Supplier No. NBP3071065UG
Add to Cart
Add to Cart

Recombinant Protein

Biological activity measured in a cell proliferation assay using human peripheral blood monocytes. The ED50 for this effect is 1-4 μg/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 mm filtered concentrated solution in PBS (pH 7.4).
Gene ID (Entrez) 6285
Molecular Weight (g/mol) MolecularWeight-theroretical: 10.6 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1.0 EU/μg of S100B as determined by LAL method.
Gene Alias beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10 protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Gene Symbol S100B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells
Purity or Quality Grade >95%, by SDS-PAGE
Protein S100B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.