Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Semaphorin 3F Fc Chimera Protein

Catalog No. 9878S3025
Click to view available options
Quantity:
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human Neuropilin-2 Fc Chimera (Catalog # 2215-N2 ) is immobilized at 2 μg/mL, 100 μL/well, it binds Recombinant Human Semaphorin 3F Fc Chimera. The concentration of Recombinant Human Semaphorin 3F Fc Chimera that produces 50% of the optimal binding response is 0.1-0.6 μg/mL.

Specifications

Accession Number Q13275-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose.
Gene ID (Entrez) 6405
Molecular Weight (g/mol) M.W. (Observed): 91-125 kDa, reducing conditions; M.W. (theoretical): 111 kDa
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human Semaphorin 3 F protein Human Semaphorin 3 F (Ser19-Gln772) (Leu503Met, Arg585Ala, Arg586Ala, Arg651Ala) (N-terminus) Accession # Q13275-1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity Semaphorin 3F
Species Human
Reconstitution Reconstitute at 500 μg/mL aq. soln..
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Semaphorin 3 F
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.