Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Semaphorin 7A Isoform 1 Protein
SDP

Catalog No. NBP26197750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26197750 50 μg
NBP26197710 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26197750 Supplier Novus Biologicals™ Supplier No. NBP26197750UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal hIgG-His-tag and corresponding to the amino acids 45-648 of Human Semaphorin 7A The Recombinant Human Semaphorin 7A Isoform 1 Protein is derived from Baculovirus. The Recombinant Human Semaphorin 7A Isoform 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 8482
Molecular Weight (g/mol) 95.7 kDa
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol SEMA7A
Cross Reactivity Semaphorin 7A
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.