Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SF2 Protein
SDP

Catalog No. p-7111073
Click to view available options
Quantity:
250 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-248 of Human SF2 The Recombinant Human SF2 Protein is derived from E. coli. The Recombinant Human SF2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 100mM KCl, 1mM DTT, 0.2mM EDTA, 40% Glycerol
Gene ID (Entrez) 6426
Molecular Weight (g/mol) 28.5 kDa
Name Recombinant Human SF2 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias Alternative-splicing factor 1, ASFpre-mRNA-splicing factor SF2, P33 subunit, MGC5228, serine/arginine-rich splicing factor 1, SF2FLJ53078, SF2p33, SFRS1splicing factor 2, Splicing factor, arginine/serine-rich 1ASF-1, SR splicing factor 1, SRp30a
Gene Symbol SRSF1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.