Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Siglec-3/CD33 Fc Avi-Tag Protein
Click to view available options
Quantity:
50 μg
Description
Measured by the ability of the immobilized protein to support the adhesion of human red blood cells. Kelm, S. et al. (1994) Current Biology 4:965. The ED50 for this effect is 0.2-1.4 μg/mL. Measured by its binding ability in a functional ELISA. When Human Siglec-3/CD33 (6C5/2) Antibody (Catalog # MAB1137 ) is immobilized at 0.5 μg/mL (100 μL/well), Biotinylated Recombinant Human Siglec-3/CD33 Fc Chimera (Catalog # AVI10467) binds with an ED50 of 0.15-1.2 μg/mL.
Specifications
Specifications
Accession Number | P20138.2 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Gene ID (Entrez) | 945 |
Molecular Weight (g/mol) | M.W. (Observed): 67-78 kDa, under reducing conditions; M.W. (theoretical): 15 kDa |
Quantity | 50 μg |
Source | Human embryonic kidney cell, HEK293-derived human Siglec-3/CD33 protein Human Siglec-3/CD33 (Met17-His259) (N-terminus) Accession # P20138.2 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Conjugate | Biotin |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction