Learn More
Novus Biologicals™ Recombinant Human SIRP gamma/CD172g hIgG-His Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Concentration | 0.5 mg/ml |
| For Use With (Application) | Bioactivity, Functional Assay |
| Formulation | PBS (pH 7.4), 20% glycerol |
| Gene ID (Entrez) | 55423 |
| Molecular Weight (g/mol) | 64 kDa |
| Quantity | 0.25 mg |
| Storage Requirements | Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
| Gene Alias | bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma |
| Gene Symbol | SIRPG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.