Learn More
Description
- Secreted Ly-6/uPAR domain-containing protein 2 (SLURP2) is expressed primarily in epithelial and immune cells and regulates growth and differentiation of epithelial cells.
- Mature SLURP2 is a secreted protein which is comprised of 75 amino acids. Human SLURP2 shares 65% and 25% aa identity with mouse and rat SLURP2, respectively.
- SLURP2 is demonstrated to lessen the tumorigenic activity of nitrosamines on Het-1A cells, reduce inflammation on human intestinal epithelial cells and Immunocytes such as CEM and U937.
- SLURP2 can interact with alpha 3, alpha 4, alpha 5, alpha 7, beta 2, beta 4, and possibly alpha 6 nicotinic acetylcholine receptors (nAChRs) as well as M1 and M3 muscarinic acetylcholine receptors.
- In addition, SLURP2 controls the proliferation of keratinocytes and epithelial cancer cells via nAChRs on the cell surface.
- It has been suggested SLURP2 may be involved in the pathophysiology of psoriasis through its role in keratinocyte hyper-proliferation and/or T cell differentiation/activation.
Specifications
Specifications
| Accession Number | P0DP57 |
| Molecular Weight (g/mol) | 35 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human SLURP2 protein MDHuman IgG1 (Pro100-Lys330)IEGRHuman SLURP2 (Ile23-Asp97) Accession # P0DP57N-terminus C-terminus |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Reconstitution | Reconstitute at 500 μg/mL in PBS |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
