Learn More
Novus Biologicals Recombinant Human SOD1/Cu-Zn SOD Pre-formed Fibrils Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
| Formulation | PB pH 7.4, 5 mM EDTA, 50 mM DTT |
| Gene ID (Entrez) | 6647 |
| Quantity | 5 x 100 μg Vials |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic |
| Gene Symbol | SOD1 |
| Research Category | Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | SOD1/Cu-Zn SOD |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.