Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SOD1/Cu-Zn SOD Protein
SDP

Catalog No. NB170844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170844 0.02 mg
NBC118544 0.1mg
NB170845 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB170844 Supplier Novus Biologicals™ Supplier No. NBC1185440.02MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

Specific activity is > 500 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.5 at 25C.

Specifications

Accession Number P22309
For Use With (Application) SDS-PAGE, Bioactivity
Formulation 20 mM Tris buffer (pH 7.5), 10% glycerol
Gene ID (Entrez) 6647
Molecular Weight (g/mol) 16 kDa
Name SOD1/Cu-Zn SOD Protein
Purification Method Protein
Quantity 0.02 mg
Source E. Coli
Immunogen SOD1, 1-154 aa, Human, Recombinant, E.coli (NP_000454).
Storage Requirements -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.