Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Sodium Potassium ATPase Beta 1 Protein

Click to view available options
Quantity:
0.5 mg
100 μg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 63 - 303 of Human Sodium Potassium ATPase Beta 1 The Recombinant Human Sodium Potassium ATPase Beta 1 Protein is derived from E. coli. The Recombinant Human Sodium Potassium ATPase Beta 1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
For Use With (Application) | SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 0.4M urea |
Gene ID (Entrez) | 481 |
Molecular Weight (g/mol) | 30.4kDa |
Name | Human Sodium Potassium ATPase Beta 1 Protein |
Purification Method | >90%, by SDS-PAGE |
Quantity | 100 μg |
Source | E.Coli |
Immunogen | MGSSHHHHHH SSGLVPRGSH MGSEFKPTYQ DRVAPPGLTQ IPQIQKTEIS FRPNDPKSYE AYVLNIVRFL EKYKDSAQRD DMIFEDCGDV PSEPKERGDF NHERGERKVC RFKLEWLGNC SGLNDETYGY KEGKPCIIIK LNRVLGFKPK PPKNESLETY PVMKYNPNVL PVQCTGKRDE DKDKVGNVEY FGLGNSPGFP LQYYPYYGKL LQPKYLQPLL AVQFTNLTMD TEIRIECKAY GENIGYSEKD RFQGRFDVKI EVKS |
Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction