Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sonic Hedgehog/Shh, N-Terminus Protein

Catalog No. 1314SH025
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Sonic Hedgehog/Shh, N-Terminus Protein is derived from E. coli. The Recombinant Human Sonic Hedgehog/Shh, N-Terminus Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) 20 kDa
Name Sonic Hedgehog/Shh N-Terminus
Quantity 25 μg
Source E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE with silver staining, under reducing conditions.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.