Learn More
Description
Specifications
Specifications
| Accession Number | NP_008960 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 50mM Tris-HCl buffer (pH7.5), 10% glycerol |
| Gene ID (Entrez) | 11075 |
| Molecular Weight (g/mol) | 16.4kDa |
| Name | Stathmin-2/STMN2 Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MDMEVKQINK RASGQAFELI LKPPSPISEA PRTLASPKKK DLSLEEIQKK LEAAEERRKS QEAQVLKQLA EKREHEREVL QKALEENNNF SKMAEEKLIL KMEQIKENRE ANLAAIIERL QEKERHAAEV RRNKELQVEL SG |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
