Learn More
Novus Biologicals Recombinant Human Tau 441 Active, P301S Mutant, Pre-formed Fibrils Protein [ATTO 488]
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
| Formulation | PBS pH 7.4 |
| Gene ID (Entrez) | 4137 |
| Quantity | 2 x 100 μg Vials |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | MAPT, microtubule-associated protein tau, MTBT1, MTBT2 |
| Gene Symbol | MAPT |
| Purity or Quality Grade | >95% |
| Protein | Tau 441 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.