Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Tau 441 Protein
SDP

Catalog No. NP2769420UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2769420UG 200 μg
NP2769410UG 100 μg
NP2767940UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NP2769420UG Supplier Novus Biologicals™ Supplier No. NBP276794200UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Western Blot, In vitro assay, In vivo assay, SDS-Page

An un-tagged full length Human Recombinant Tau 441 protein Pre-Formed Fibrils, Accession #: P10636

The Recombinant Human Tau 441 Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page.

Specifications

For Use With (Application) Western Blot, In vitro Assay, In vivo Assay, SDS-PAGE
Gene ID (Entrez) 4137
Molecular Weight (g/mol) 45.8 kDa
Quantity 200 μg
Source E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol MAPT
Specific Reactivity Tau 441
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 10 mM HEPES, 100 mM NaCl pH 7.4
Purity or Quality Grade 0.2 Micrometer Filtered
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.