Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Tau Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Human Tau Protein is derived from E. coli. The Recombinant Human Tau Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | NP_005901.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 4137 |
| Molecular Weight (g/mol) | 46 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived human Tau protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Specific Reactivity | Tau |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction