Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TCEB2 Protein
SDP

Catalog No. NBC122579
Click to view available options
Quantity:
0.1 mg

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-118 of Human TCEB2 The Recombinant Human TCEB2 Protein is derived from E. coli. The Recombinant Human TCEB2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q15370
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1mM PMSF, 20% glycerol
Gene ID (Entrez) 6923
Molecular Weight (g/mol) 13.3kDa
Name TCEB2 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Elongin B, 1-118aa, Human, Recombinant, E.coli (NP_009039).
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol ELOB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.