Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TDO2 Protein

Catalog No. 9768TD020
Click to view available options
Quantity:
20 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Tryptophan 2,3-dioxygenase

Measured by its ability to oxidize L-tryptophan to N-formyl-kynurenine. The specific activity is >650 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P48775
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 6999
Molecular Weight (g/mol) M.W. (Observed): 40 kDa and 78 kDa, reducing conditions; M.W. (theoretical): 45 kDa
Name chky, TDO, TDOTO, TO, TPH2EC 1.13.11.11, TRPO, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan Oxygenase, Tryptophan Pyrrolase, Tryptophanase
Quantity 20 μg
Source E. coli-derived human TDO2 protein Leu18-Phe388 with an N-terminal Met and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TDO2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.