Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TGF-beta 1 Protein
SDP

Catalog No. NB002039
Click to view available options
Quantity:
5 μg

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein for Human TGF-beta 1 The Recombinant Human TGF-beta 1 Protein is derived from CHO. The Recombinant Human TGF-beta 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from Sodium citrate buffer (pH 3.5).
Gene ID (Entrez) 7040
Molecular Weight (g/mol) 25.0 kDa
Quantity 5 μg
Source CHO
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Gene Symbol TGFB1
Reconstitution Reconstitution in H2O to 50 μg/mL will yield a solution of 1 mM citrate buffer (pH 3.5). The solution can then be stored at 4°C for 1 month. Further dilutions should be made in PBS containing 2 mg/mL albumin and stored at -20°C.
Buffer Lyophilized from Sodium citrate buffer (pH 3.5).
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.